missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One VACUETTE™ Z Serum Blood Collection Tubes

Product Code. 13068662
Change view
Click to view available options
Capacity (Metric):
2 mL
Closure Color:
Red with White Ring
Unit Size:
Pack of 1200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Color Unit Size
13068662 2 mL Red with White Ring Pack of 1200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13068662 Supplier Greiner Bio-One Supplier No. 454096
  • ONLINE EXCLUSIVE PRICE
    £217.00
    £198.48 / Pack of 1200
    Save £18.52
    8% Off
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VACUETTE™ Z Serum Blood Collection Tubes are used in determinations for clinical chemistry, immunology and serology. Available in Serum Clot Activator, Serum Sep Clot Activator and Serum Fast Separator. The inner wall of the tube is coated with silica particles which activate clotting when the tubes are turned.

  • Tubes are supplied in packs of 50 pieces
  • For venus blood collection
  • Primarily used for drawing blood for clinical chemistry, immunology and serology testing
  • Specially coated with microscopic silica particles, which activate the coagulation process
  • Made out of virtually unbreakable PET plastic
  • Provides improved safety and hygiene during specimen collection procedures with the advantage of being as clear as glass
  • Available in: Serum Clot Activator, Serum Sep Clot Activator and Serum Fast Separator
  • CE marked
TRUSTED_SUSTAINABILITY

Specifications

Product Type Blood Collection Tube
Additive Clot Activator
Sterility Sterile
Closure Type Premium Safety Twist Cap
Capacity (Metric) 2 mL
For Use With (Equipment) VACUETTE™ Blood Collection Needles
Closure Color Red with White Ring
Tube Size 13 x 75 mm
Quantity Case of 24
For Use With (Application) Blood Collection
Length (Metric) 75 mm
Diameter (Metric) Outer 13 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.