missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Serum Bottles
Shop All Labconco Corporation Products
Click to view available options
Capacity (Metric):
2 mL
3 mL
10 mL
20 mL
50 mL
100 mL
125 mL
Unit Size:
Pack of 100
Description
- Uniform thin wall construction ensures even freezing and drying
- Ideal containers for use in the FreeZone Stoppering Tray Dryer
- May be connected to valve ports on drying chambers and manifolds
- Supplied in packages of 100
Specifications
Specifications
| Capacity (Metric) | 100 mL |
| Color | Clear |
| Material | Glass |
| No. per Pack | 100/PK |
| Sterility | Non-sterile |
| Product Type | Serum Bottle |
| Closure Size | 20 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction