missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Serum Bottles

Product Code. 15123065 Shop All Labconco Corporation Products
Change view
Click to view available options
Capacity (Metric):
2 mL
3 mL
10 mL
20 mL
50 mL
100 mL
125 mL
Unit Size:
Pack of 100
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15123065 20 mL Pack of 100
15163065 2 mL Pack of 100
15153065 3 mL Pack of 100
15113065 10 mL Pack of 100
15133065 50 mL Pack of 100
15143065 100 mL Pack of 100
15173065 125 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 15123065 Supplier Labconco™ Supplier No. 7573410
  • ONLINE EXCLUSIVE PRICE
    £300.00
    £293.03 / Pack of 100
    Save £6.97
    2% Off
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Perfect for long term storage of freeze dried samples. Labconco Serum Bottles are specifically designed for lyophilization applications.

  • Uniform thin wall construction ensures even freezing and drying
  • Ideal containers for use in the FreeZone Stoppering Tray Dryer
  • May be connected to valve ports on drying chambers and manifolds
  • Supplied in packages of 100

Specifications

Capacity (Metric) 20 mL
Color Clear
Material Glass
Quantity 100/PK
Sterility Non-sterile
Product Type Serum Bottle
Closure Size 20 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.