missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Serum Amyloid P Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA524171
This item is not returnable.
View return policy
Description
Serum Amyloid P Polyclonal Antibody for Western Blot, IHC (P)
The protein encoded by this gene is a glycoprotein, belonging to the pentraxin family of proteins, which has a characteristic pentameric organization. These family members have considerable sequence homology which is thought to be the result of gene duplication. The binding of the encoded protein to proteins in the pathological amyloid cross-beta fold suggests its possible role as a chaperone. This protein is also thought to control the degradation of chromatin. It has been demonstrated that this protein binds to apoptotic cells at an early stage, which raises the possibility that it is involved in dealing with apoptotic cells in vivo.
Specifications
| Serum Amyloid P | |
| Polyclonal | |
| Unconjugated | |
| APCS | |
| 9.5S alpha-1-glycoprotein; amyloid P component, serum; APCS; APCS amyloid P-component serum protein; epididymis secretory sperm binding protein Li 92n; HEL-S-92n; MGC88159; Pentaxin related; pentaxin-related; Pentraxin 2; pentraxin-2; pentraxin-related; PTX 2; PTX2; SAMP; Sap; Serum amyloid P-component; Serum amyloid P-component(1-203) | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| 325 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P02743 | |
| APCS | |
| KLH conjugated synthetic peptide between 151-178 amino acids from the C-terminal region of human APCS. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction