Learn More
Invitrogen™ Serum Amyloid P Monoclonal Antibody (273912)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA523975
Description
In Western blots, 100% cross-reactivity with recombinant human (rh) Pentraxin 2 and 25% cross-reactivity with recombinant mouse (rm) Pentraxin 2 is observed. In direct ELISAs, no cross-reactivity with rhPentraxin 2 or rmPentraxin 2 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
The protein encoded by this gene is a glycoprotein, belonging to the pentraxin family of proteins, which has a characteristic pentameric organization. These family members have considerable sequence homology which is thought to be the result of gene duplication. The binding of the encoded protein to proteins in the pathological amyloid cross-beta fold suggests its possible role as a chaperone. This protein is also thought to control the degradation of chromatin. It has been demonstrated that this protein binds to apoptotic cells at an early stage, which raises the possibility that it is involved in dealing with apoptotic cells in vivo.
Specifications
| Serum Amyloid P | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P23680 | |
| APCS | |
| Mouse myeloma cell line NS0-derived recombinant rat Pentraxin2/SAP Gln21-Ser228 (predicted). | |
| 100 μg | |
| Primary | |
| Rat | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 273912 | |
| Unconjugated | |
| APCS | |
| 9.5S alpha-1-glycoprotein; amyloid P component, serum; APCS; APCS amyloid P-component serum protein; epididymis secretory sperm binding protein Li 92n; HEL-S-92n; MGC88159; Pentaxin related; pentaxin-related; Pentraxin 2; pentraxin-2; pentraxin-related; PTX 2; PTX2; SAMP; Sap; Serum amyloid P-component; Serum amyloid P-component(1-203) | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 29339 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.