missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serum Amyloid A4 Rabbit anti-Human, Clone: 14, Novus Biologicals™
Rabbit Monoclonal Antibody
£286.00 - £429.00
Specifications
| Antigen | Serum Amyloid A4 |
|---|---|
| Clone | 14 |
| Dilution | Sandwich ELISA Detection 1:1000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18279107
|
Novus Biologicals
NBP2-89368-50UL |
50 μg |
£286.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18263637
|
Novus Biologicals
NBP2-89368-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Serum Amyloid A4 Monoclonal specifically detects Serum Amyloid A4 in Human samples. It is validated for Sandwich ELISA Detection.Specifications
| Serum Amyloid A4 | |
| Sandwich ELISA Detection 1:1000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 6291 | |
| IgG | |
| Protein A purified |
| 14 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| C-SAACSAAConstitutively expressed serum amyloid A protein, serum amyloid A-4 protein, serum amyloid A4, constitutive | |
| Recombinant Human Serum Amyloid A4 Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title