missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serum Amyloid A4 Antibody (014), Alexa Fluor™ 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89368AF488
This item is not returnable.
View return policy
Description
Serum Amyloid A4 Monoclonal specifically detects Serum Amyloid A4 in Human samples. It is validated for Sandwich ELISA Detection.
Specifications
| Serum Amyloid A4 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 14 | |
| Sandwich ELISA Detection | |
| C-SAACSAAConstitutively expressed serum amyloid A protein, serum amyloid A-4 protein, serum amyloid A4, constitutive | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Serum Amyloid A4 (P35542; Glu19-Tyr130). | |
| 0.1 mL | |
| Immunology | |
| 6291 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction