missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serum Amyloid A1 Antibody (SAA15cc), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-73077
This item is not returnable.
View return policy
Description
Serum Amyloid A1 Monoclonal specifically detects Serum Amyloid A1 in Human, Canine samples. It is validated for Western Blot, ELISA.
Specifications
| Serum Amyloid A1 | |
| Monoclonal | |
| Unconjugated | |
| P02735 | |
| SAA1 | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Human SAA. | |
| 0.2 mg | |
| Alzheimers Research, Cancer, Neuroscience | |
| 6288 | |
| Human, Canine | |
| Purified |
| Western Blot, ELISA | |
| SAA15cc | |
| Western Blot, ELISA | |
| MGC111216, PIG4, SAA, SAA2, serum amyloid A protein, serum amyloid A1, TP53I4, tumor protein p53 inducible protein 4 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human recombinant SAA. Also cross-reacts with canine SAA. | |
| Store at 4C. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction