Learn More
Serum Amyloid A1 Antibody (CL9175) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21169C
Product Code. 30307668
  • £558.00 / 0.10mL
Description
Serum Amyloid A1 Monoclonal antibody specifically detects Serum Amyloid A1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Serum Amyloid A1
Monoclonal
DyLight 650
MGC111216, PIG4, SAA, SAA2, serum amyloid A protein, serum amyloid A1, TP53I4, tumor protein p53 inducible protein 4
This antibody was developed using a synthetic peptide derived from P0DJI8, with the exact immunogen sequence remaining proprietary.
0.1 mL
Alzheimers Research, Biologically Active Proteins, Cancer, Neuroscience
6288
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL9175
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified