missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serum Amyloid A1 Antibody (CL9175) [Alexa Fluor™ 700], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21169AF700
This item is not returnable.
View return policy
Description
Serum Amyloid A1 Monoclonal antibody specifically detects Serum Amyloid A1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Serum Amyloid A1 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| MGC111216, PIG4, SAA, SAA2, serum amyloid A protein, serum amyloid A1, TP53I4, tumor protein p53 inducible protein 4 | |
| This antibody was developed using a synthetic peptide derived from P0DJI8, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Alzheimers Research, Biologically Active Proteins, Cancer, Neuroscience | |
| 6288 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL9175 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction