missing translation for 'onlineSavingsMsg'
Learn More

Serum Amyloid A1 Antibody (370820) [HRP], Novus Biologicals™

Product Code. 30737448 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30737448 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737448 Supplier Novus Biologicals Supplier No. FAB3019H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Serum Amyloid A1 Monoclonal antibody specifically detects Serum Amyloid A1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serum Amyloid A1
Applications Western Blot
Classification Monoclonal
Clone 370820
Conjugate HRP
Formulation PBS
Gene Alias MGC111216, PIG4, SAA, SAA2, serum amyloid A protein, serum amyloid A1, TP53I4, tumor protein p53 inducible protein 4
Host Species Rat
Immunogen E. coli-derived recombinant human SAA1, Arg19-Tyr122, Accession # P02735
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Biologically Active Proteins, Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6288
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.