missing translation for 'onlineSavingsMsg'
Learn More

Serum Amyloid A1/A2 Rabbit anti-Human, Clone: SAA/2868R, Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-07767-100ug

 View more versions of this product

Product Code. 18661868

  • £431.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Serum Amyloid A1/A2 Monoclonal specifically detects Serum Amyloid A1/A2 in Human samples. It is validated for Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Serum Amyloid A1/A2
Monoclonal
0.2 mg/mL
Immunohistochemistry-Paraffin 1 to 2 μg/mL
Rabbit
Protein A or G purified
RUO
6288
Store at 4°C.
IgG
Immunohistochemistry (Paraffin)
SAA/2868R
Unconjugated
10 mM PBS with 0.05% BSA
Recombinant full-length human Serum Amyloid A1/A2 (SAA) protein
100 μg
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.