missing translation for 'onlineSavingsMsg'
Learn More

Serum Amyloid A1/A2 Antibody (SR1117), Novus Biologicals™

Rabbit Monoclonal Antibody

£265.00 - £358.00

Specifications

Antigen Serum Amyloid A1/A2
Clone SR1117
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18616124
View Documents
Novus Biologicals
NBP3-21727-100ul
100 μg
£358.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18630754
View Documents
Novus Biologicals
NBP3-21727-50ul
50 μg
£265.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Serum Amyloid A1/A2 Monoclonal antibody specifically detects Serum Amyloid A1/A2 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

Serum Amyloid A1/A2
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200
Monoclonal
Purified
RUO
PBS (pH 7.4), 150 mM NaCl, 50% Glycerol
6288
IgG
Affinity purified
SR1117
Western Blot, Immunohistochemistry
Unconjugated
Rabbit
Human
PIG4;SAA;SAA1;SAA2;Serum amyloid A-1 protein;Serum amyloid A-2 protein;TP53I4
A synthesized peptide derived from human Serum Amyloid A1/A2 (Uniprot #: P0DJI8)
Primary
Store at -20°C. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.