missing translation for 'onlineSavingsMsg'
Learn More

Serum Amyloid A1/A2 Antibody (SAA/2868R), Alexa Fluor™ 750, Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-08413AF750

Product Code. 18518251

  • £510.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Serum Amyloid A1/A2 Monoclonal antibody specifically detects Serum Amyloid A1/A2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Serum Amyloid A1/A2
Monoclonal
Alexa Fluor 750
PIG4;SAA;SAA1;SAA2;Serum amyloid A-1 protein;Serum amyloid A-2 protein;TP53I4
Recombinant full-length human Serum Amyloid A1/A2 (SAA) protein
100 μg
Primary
Human
Purified
Immunohistochemistry (Paraffin)
SAA/2868R
50mM Sodium Borate
Rabbit
Protein A or G purified
RUO
6288
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.