missing translation for 'onlineSavingsMsg'
Learn More

Serum Amyloid A1/A2 Antibody (834835R) [HRP], Novus Biologicals™

Product Code. 30733667 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30733667 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733667 Supplier Novus Biologicals Supplier No. FAB2948H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Serum Amyloid A1/A2 Monoclonal antibody specifically detects Serum Amyloid A1/A2 in Mouse samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serum Amyloid A1/A2
Applications ELISA
Classification Monoclonal
Clone 834835R
Conjugate HRP
Formulation PBS
Gene Alias PIG4;SAA;SAA1;SAA2;Serum amyloid A-1 protein;Serum amyloid A-2 protein;TP53I4
Host Species Rat
Immunogen E. coli-derived recombinant mouse Serum Amyloid A1/A2, Gly20-Tyr122, Accession # P05366
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6288
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.