missing translation for 'onlineSavingsMsg'
Learn More

Sertraline hydrochloride, MedChemExpress

Product Code. 16424277
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16424277

Brand: Medchem Express HYB0176A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sertraline hydrochloride is an antidepressant of the selective serotonin reuptake inhibitor (SSRI) class. Sertraline hydrochloride is researched for a number of diseases, such as major depressive disorder and obsessive.

Sertraline hydrochloride is an antidepressant of the selective serotonin reuptake inhibitor (SSRI) class. Sertraline hydrochloride is researched for a number of diseases, such as major depressive disorder and obsessive.

Specifications

Chemical Name or Material Sertraline hydrochloride
CAS 79559-97-0
Color White
Purity Grade Notes Research
Molecular Formula C17H18Cl3N
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (145.90 mM; Need ultrasonic) ∣H2O : 2.86 mg/mL (8.35 mM; ultrasonic and warming and heat to 60°C)
SMILES CN[C@H]1CC[C@@H](C2=CC=C(Cl)C(Cl)=C2)C3=C1C=CC=C3.Cl
Molecular Weight (g/mol) 342.69
Formula Weight 342.69
Percent Purity 99.38%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.