missing translation for 'onlineSavingsMsg'
Learn More

Sertraline hydrochloride, MedChemExpress

Product Code. 16490798
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490798 5 mg 5mg
16424277 50 mg 50mg
16418388 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16490798 Supplier Medchem Express Supplier No. HYB0176A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sertraline hydrochloride is an antidepressant of the selective serotonin reuptake inhibitor (SSRI) class. Sertraline hydrochloride is researched for a number of diseases, such as major depressive disorder and obsessive.

Sertraline hydrochloride is an antidepressant of the selective serotonin reuptake inhibitor (SSRI) class. Sertraline hydrochloride is researched for a number of diseases, such as major depressive disorder and obsessive.

Specifications

Chemical Name or Material Sertraline hydrochloride
CAS 79559-97-0
Color White
Purity Grade Notes Research
Molecular Formula C17H18Cl3N
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (145.90 mM; Need ultrasonic) ∣H2O : 2.86 mg/mL (8.35 mM; ultrasonic and warming and heat to 60°C)
SMILES CN[C@H]1CC[C@@H](C2=CC=C(Cl)C(Cl)=C2)C3=C1C=CC=C3.Cl
Molecular Weight (g/mol) 342.69
Formula Weight 342.69
Percent Purity 99.38%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.