missing translation for 'onlineSavingsMsg'
Learn More

Sertraline Carbamoyl Glucuronide, TRC

Product Code. 30428363
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30428363 5 mg 5mg
30417093 1 mg -
30420979 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30428363 Supplier Toronto Research Chemicals Supplier No. TRCS2799905MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119733-44-7
Molecular Formula C24 H25 Cl2 N O8
Molecular Weight (g/mol) 526.36
IUPAC Name (2S,3S,4S,5R,6S)-6-[[(1S,4S)-4-(3,4-dichlorophenyl)-1,2,3,4-tetrahydronaphthalen-1-yl]-methylcarbamoyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
SMILES CN([C@H]1CC[C@@H](c2ccc(Cl)c(Cl)c2)c3ccccc13)C(=O)O[C@@H]4O[C@@H]([C@@H](O)[C@H](O)[C@H]4O)C(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C24H25Cl2NO8/c1-27(24(33)35-23-20(30)18(28)19(29)21(34-23)22(31)32)17-9-7-12(13-4-2-3-5-14(13)17)11-6-8-15(25)16(26)10-11/h2-6,8,10,12,17-21,23,28-30H,7,9H2,1H3,(H,31,32)/t12-,17-,18-,19-,20+,21-,23-/m0/s1
Formula Weight 525.0957
Chemical Name or Material Sertraline Carbamoyl Glucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.