missing translation for 'onlineSavingsMsg'
Learn More

Sertindole, MedChemExpress

Product Code. 16462227
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16462227

Brand: Medchem Express HY14543

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sertindole, a neuroleptic, is one of the newer antipsychotic medications available.

Sertindole, a neuroleptic, is one of the newer antipsychotic medications available.

Specifications

Chemical Name or Material Sertindole
CAS 106516-24-9
Color White
Purity Grade Notes Research
Molecular Formula C24H26ClFN4O
Quantity 50 mg
Synonym Lu 23-174
Solubility Information DMSO : 25 mg/mL (56.70 mM; Need ultrasonic)
SMILES FC1=CC=C(N2C3=CC=C(Cl)C=C3C(C4CCN(CCN5CCNC5=O)CC4)=C2)C=C1
Molecular Weight (g/mol) 440.94
Formula Weight 440.94
Percent Purity 95.04%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.