missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SERTAD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543700
This item is not returnable.
View return policy
Description
Peptide sequence: LQRQTIFNIS LMKLYNHRPL TEPSLQKTVL INNMLRRIQE ELKQEGSLRP Sequence homology: Cow: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 77%.
Acts at E2F-responsive promoters as coregulator to integrate signals provided by PHD- and/or bromodomain-containing transcription factors. May act as coactivator as well as corepressor of E2F1-TFDP1 and E2F4-TFDP1 complexes on E2F consensus binding sites, which would activate or inhibit E2F-target genes expression. Modulates fat storage by down-regulating the expression of key genes involved in adipocyte lipolysis, thermogenesis and oxidative metabolism. [UniProt].
Specifications
| SERTAD2 | |
| Polyclonal | |
| Unconjugated | |
| SERTAD2 | |
| AB041541; AU021878; Kiaa0127; MGC126688; MGC126690; mKIAA0127; MNCb-1504; PHD zinc finger- and bromodomain-interacting protein 2; Sei2; Sei-2; SERTA domain containing 2; SERTA domain-containing protein 2; SERTAD2; Transcriptional regulator interacting with the PHD-bromodomain 2; transcriptional regulator interacting with the PHS-bromodomain 2; TRIPBR2; TRIP-Br2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 9792 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q14140 | |
| SERTAD2 | |
| Synthetic peptide directed towards the N-terminal of human SERTAD2 (aa 35-84). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction