missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sertad1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94785-0.02ml
This item is not returnable.
View return policy
Description
Sertad1 Polyclonal antibody specifically detects Sertad1 in Rat samples. It is validated for Western Blot
Specifications
| Sertad1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| SEI-1, SEI1CDK4-binding protein p34SEI1, SERTA domain containing 1, SERTA domain-containing protein 1, Transcriptional regulator interacting with the PHD-bromodomain 1, TRIP-Br1CDK4-binding protein p34SEI | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-236 of human SERTAD1 (NP_037508.2). MLSKGLKRKREEEEEKEPLAVDSWWLDPGHTAVAQAPPAVASSSLFDLSVLKLHHSLQQSEPDLRHLVLVVNTLRRIQASMAPAAALPPVPSPPAAPSVADNLLASSDAALSASMASLLEDLSHIEGLSQAPQPLADEGPPGRSIGGAAPSLGALDLLGPATGCLLDDGLEGLFEDIDTSMYDNELWAPASEGLKPGPEDGPGKEEAPELDEAELDYLMDVLVGTQALERPPGPGR | |
| 0.02 mL | |
| Cell Cycle and Replication | |
| 29950 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction