missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sertaconazole Nitrate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Sertaconazole Nitrate
Specifications
Specifications
| Chemical Name or Material | Sertaconazole Nitrate |
| CAS | 99592-39-9 |
| Analyte or Component Names | Sertaconazole Nitrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H15 Cl3 N2 O S . H N O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Sertaconazole nitrate, (RS)-1-[2-[(7-Chloro-1-benzothiophen-3-yl)methoxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole nitrate, 1H-Imidazole, 1-[2-[(7-chlorobenzo[b]thien-3-yl)methoxy]-2-(2,4-dichlorophenyl)ethyl]-, nitrate (1:1), 1H-Imidazole, 1-[2-[(7-chlorobenzo[b]thien-3-yl)methoxy]-2-(2,4-dichlorophenyl)ethyl]-, mononitrate (9CI), Dermofix, Dermovit, Ertaczo, FI 7056, Sertaconazole mononitrate, Zalain |
| InChI Formula | InChI=1S/C20H15Cl3N2OS.HNO3/c21-14-4-5-16(18(23)8-14)19(9-25-7-6-24-12-25)26-10-13-11-27-20-15(13)2-1-3-17(20)22;2-1(3)4/h1-8,11-12,19H,9-10H2;(H,2,3,4) |
| SMILES | O[N+](=O)[O-].Clc1ccc(C(Cn2ccnc2)OCc3csc4c(Cl)cccc34)c(Cl)c1 |
| IUPAC Name | 1-[2-[(7-chloro-1-benzothiophen-3-yl)methoxy]-2-(2,4-dichlorophenyl)ethyl]imidazole;nitric acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction