missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sertaconazole nitrate, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
Sertaconazole nitrate (FI7056) is a broad-spectrum topical antifungal agent, exhibits anti-inflammatory activity via activation of a p38-COX-2-PGE2 pathway. Sertaconazole nitrate is also a microtubule inhibitor, shows antiproliferative effect, induces apoptosis and autophagy, and can also inhibit the migration of cells.
Specifications
Specifications
| Chemical Name or Material | Sertaconazole nitrate |
| Physical Form | Solid |
| CAS | 99592-39-9 |
| Color | Off-White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C20H16Cl3N3O4S |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | FI7056 |
| Solubility Information | DMSO : ≥ 100 mg/mL (199.69 mM) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction