missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SERPINI2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116265
This item is not returnable.
View return policy
Description
SERPINI2 Polyclonal Antibody for Western Blot
The protein encoded by this gene is a member of the serine protease inhibitor superfamily made up of proteins which play central roles in the regulation of a wide variety of physiological processes, including coagulation, fibrinolysis, development, malignancy and inflammation. The gene product may have a role in a growth-control, possibly growth-suppressing pathway and, when impaired, may be involved in pancreatic carcinogenesis. The protein is a member of the plasminogen activator inhibitor-1 family, a subset of the serpin superfamily whose members act as tissue-specific tPA inhibitors. Two alternatively spliced transcript variants encoding distinct protein isoforms have been found for this gene.
Specifications
| SERPINI2 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O75830 | |
| SERPINI2 | |
| KLH conjugated synthetic peptide between 375-401 amino acids from the C-terminal region of human SERPINI2. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| SERPINI2 | |
| MEPI; Myoepithelium-derived serine protease inhibitor; neuroserpin; PANCPIN; Pancreas-specific protein TSA2004; pancreas-specific serpin; peptidase inhibitor 14; PI14; PI-14; protease inhibitor 14; serine (or cysteine) peptidase inhibitor, clade I, member 2; serine (or cysteine) proteinase inhibitor, clade I (neuroserpin), member 2; serine (or cysteine) proteinase inhibitor, clade I (pancpin), member 2; serine protease inhibitor 14; serpin family I member 2; serpin I2; serpin peptidase inhibitor, clade I (pancpin), member 2; serpin peptidase inhibitor, clade I, member 2; SERPIN12; SERPINI2; Spi14; TSA2004 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 5276 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction