Learn More
Invitrogen™ SERPINF2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547142
Description
In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant mouse (rm) Serpin F2 is observed and less than 1% cross-reactivity with recombinant human (rh) Serpin A1, rhSerpin A3, rhSerpin A4, rhSerpin A5, rmSerpin C1, and rmSerpin D1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes a member of the serpin family of serine protease inhibitors. The protein is a major inhibitor of plasmin, which degrades fibrin and various other proteins. Consequently, the proper function of this gene has a major role in regulating the blood clotting pathway. Mutations in this gene result in alpha-2-plasmin inhibitor deficiency, which is characterized by severe hemorrhagic diathesis. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| SERPINF2 | |
| Polyclonal | |
| Unconjugated | |
| SERPINF2 | |
| A2AP; AAP; AI747498; alpha 2 antiplasmin; alpha-2-antiplasmin; Alpha-2-AP; alpha-2-PI; Alpha-2-plasmin inhibitor; API; pigment epithelium derived factor; plasmin inhibitor alpha 2; Pli; serine (or cysteine) peptidase inhibitor, clade F, member 2; serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 2; serine (or cysteine) proteinase inhibitor, clade F, member 2; Serpimf2; serpin F2; serpin family F member 2; serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 2; Serpinf2 | |
| Goat | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5345 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunoprecipitation, Neutralization, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P08697 | |
| SERPINF2 | |
| Mouse myeloma cell line NS0-derived recombinant human Serpin F2/alpha 2-Antiplasmin Met28-Lys491. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.