Learn More
Invitrogen™ SERPINE2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532163
Description
Recommended positive controls: HeLa, HepG2, mouse brain. Predicted reactivity: Mouse (87%), Rat (87%), Pig (94%), Chicken (82%), Bovine (93%). PA5-32163 detects SERPINE2 protein in formalin-fixed paraffin-embedded equine endometrial tissue. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the serpin family of proteins, a group of proteins that inhibit serine proteases. Thrombin, urokinase, plasmin and trypsin are among the proteases that this family member can inhibit. This gene is a susceptibility gene for chronic obstructive pulmonary disease and for emphysema. Alternative splicing results in multiple transcript variants.
Specifications
| SERPINE2 | |
| Polyclonal | |
| Unconjugated | |
| SERPINE2 | |
| B230326M24Rik; EGK_04851; GDN; GDNPF; Gdnpn1; Glia derived nexin; glia-derived nexin; Glia-derived nexin/protease nexin I; Glia-derived nexin-like protein; glial-derived neurite promoting factor; P17; PAI-1; Peptidase inhibitor 7; Pi7; PI-7; plasminogen activator inhibitor type 1; PN1; Pn-1; PNI; Protease nexin 1; protease nexin I; protease nexin PN-1; protease nexin-1; serine (or cysteine) peptidase inhibitor, clade E, member 2; serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 2; serine (or cysteine) proteinase inhibitor, clade E, member 2; serine protease inhibitor 4; serine protease-inhibitor 4; Serpin E2; serpin family E member 2; serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 2; serpin peptidase inhibitor, clade E, member 2; SERPINE 2; SERPINE1; Serpine2; Spi4; Spin4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100056744, 20720, 5270 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| P07093, Q07235 | |
| SERPINE2 | |
| Recombinant protein encompassing a sequence within the center region of human SERPINE2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Horse, Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.