missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINE2 Mouse anti-Human, Clone: 1E11F12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66203-1-IG-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the serpin family of proteins, a group of proteins that inhibit serine proteases. Thrombin, urokinase, plasmin and trypsin are among the proteases that this family member can inhibit. This gene is a susceptibility gene for chronic obstructive pulmonary disease and for emphysema. Alternative splicing results in multiple transcript variants.Specifications
| SERPINE2 | |
| Monoclonal | |
| 0.87 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P07093 | |
| SERPINE2 | |
| SERPINE2 Fusion Protein Ag1830 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
| 1E11F12 | |
| Unconjugated | |
| SERPINE2 | |
| GDN, Glia derived nexin, Peptidase inhibitor 7, PI 7, PI7, PN 1, PN1, PN-1, PNI, Protease nexin 1, Protease nexin I, Serpin E2, SERPINE2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 5270 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction