missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SERPINE2 Monoclonal Antibody (OTI4E4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525933
This item is not returnable.
View return policy
Description
SERPINE2 Monoclonal Antibody for Western Blot, Flow
This gene encodes a member of the serpin family of proteins, a group of proteins that inhibit serine proteases. Thrombin, urokinase, plasmin and trypsin are among the proteases that this family member can inhibit. This gene is a susceptibility gene for chronic obstructive pulmonary disease and for emphysema. Alternative splicing results in multiple transcript variants.
Specifications
| SERPINE2 | |
| Monoclonal | |
| 4.02 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P07093, Q07235 | |
| SERPINE2 | |
| Full length human recombinant protein of SERPINE2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Canine, Monkey | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI4E4 | |
| Unconjugated | |
| SERPINE2 | |
| B230326M24Rik; EGK_04851; GDN; GDNPF; Gdnpn1; Glia derived nexin; glia-derived nexin; Glia-derived nexin/protease nexin I; Glia-derived nexin-like protein; glial-derived neurite promoting factor; P17; PAI-1; Peptidase inhibitor 7; Pi7; PI-7; plasminogen activator inhibitor type 1; PN1; Pn-1; PNI; Protease nexin 1; protease nexin I; protease nexin PN-1; protease nexin-1; serine (or cysteine) peptidase inhibitor, clade E, member 2; serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 2; serine (or cysteine) proteinase inhibitor, clade E, member 2; serine protease inhibitor 4; serine protease-inhibitor 4; Serpin E2; serpin family E member 2; serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 2; serpin peptidase inhibitor, clade E, member 2; SERPINE 2; SERPINE1; Serpine2; Spi4; Spin4 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 20720, 5270, 608930 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction