missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINB9 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14910-1-AP-20UL
This item is not returnable.
View return policy
Description
PI9 belongs to the large superfamily of serine proteinase inhibitors (serpins), which bind to and inactivate serine proteinases. These interactions are involved in many cellular processes, including coagulation, fibrinolysis, complement fixation, matrix remodeling, and apoptosis (Sprecher et al., 1995 ).Specifications
| SERPINB9 | |
| Polyclonal | |
| Unconjugated | |
| SERPINB9 | |
| CAP 3, CAP3, Cytoplasmic antiproteinase 3, Peptidase inhibitor 9, PI 9, PI9, Serpin B9, SERPINB9 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5272 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P50453 | |
| SERPINB9 | |
| SERPINB9 Fusion Protein Ag6707 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction