missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SerpinB9 Antibody (7D8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£170.00 - £464.00
Specifications
| Antigen | SerpinB9 |
|---|---|
| Clone | 7D8 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Immunohistochemistry (Frozen), Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18095254
|
Novus Biologicals
NB100-64359-0.025MG |
0.025 mg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18085784
|
Novus Biologicals
NB100-64359 |
0.1 mg |
£464.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SerpinB9 Monoclonal specifically detects SerpinB9 in Human, Mouse (Negative), Porcine (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| SerpinB9 | |
| 1.0 mg/mL | |
| Immunohistochemistry (Frozen), Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Immunology | |
| P50453 | |
| 5272 | |
| Recombinant PI-9 produced in P. pastoris. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 7D8 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse (Negative), Porcine (Negative), Human | |
| CAP-3, CAP3PI-9, Cytoplasmic antiproteinase 3, Peptidase inhibitor 9, PI9protease inhibitor 9 (ovalbumin type), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 9, serpin B9, serpin peptidase inhibitor, clade B (ovalbumin), member 9, serpin peptidase inhibitor, clade B, member 9 | |
| SERPINB9 | |
| IgG1 | |
| Protein G purified | |
| NB100-64359 is specific for human PI-9 (proteinase inhibitor 9), also known as SerpinB9, a 42kDa intracellular nucleocytoplasmic serpin expressed in cytotoxic lymphocytes (CTLs), natural killer (NK) cells, monocyte-derived dendritic cells (DCs), and to a lesser extent in B cells and myeloid cells. Granzyme B (grB) is a serine protease highly expressed by CTLs and NK cells, which is endocytosed by virus-infected and malignant target cells. The subsequent release of grB from the endocytic vesicles into the cytoplasm of the target cells, triggers grB-mediated apoptosis, through cleavage of various cytoplasmic or nuclear proteins. PI-9, up-regulated in response to grB production and degranulation, has been identified as a potent inhibitor of Granzyme B-mediated apoptosis, providing a vital self-protection mechanism against the premature apoptosis of CTLs and NK cells by grB, which may escape into the cytoplasm of the effector cells themselves. Clone 7D8 detects a band of approximately 42kDa under reducing conditions. Clone 7D8 is suitable for use in indirect Immunofluorescence (2). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title