missing translation for 'onlineSavingsMsg'
Learn More

SerpinB9 Antibody (1F5), Novus Biologicals™

Product Code. 18375919 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18375919 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18375919 Supplier Novus Biologicals Supplier No. H00005272M06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SerpinB9 Monoclonal antibody specifically detects SerpinB9 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SerpinB9
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA
Classification Monoclonal
Clone 1F5
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_004146
Gene Alias CAP-3, CAP3PI-9, Cytoplasmic antiproteinase 3, Peptidase inhibitor 9, PI9protease inhibitor 9 (ovalbumin type), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 9, serpin B9, serpin peptidase inhibitor, clade B (ovalbumin), member 9, serpin peptidase inhibitor, clade B, member 9
Host Species Mouse
Immunogen SERPINB9 (NP_004146, 279 a.a. ~ 375 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DMESVLRHLGIVDAFQQGKADLSAMSAERDLCLSKFVHKSFVEVNEEGTEAAAASSCFVVAECCMESGPRFCADHPFLFFIRHNRANSILFCGRFSS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 5272
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.