missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINB8 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13102-2-AP-150UL
This item is not returnable.
View return policy
Description
The superfamily of high molecular weight serine proteinase inhibitors. A critical portion of the molecule, the reactive center loop connects beta sheets A and C. Protease inhibitor-8 (PI8; SERPINB8) is a member of the ov-serpin subfamily, which, relative to the archetypal serpin PI1 (MIM 107400), is characterized by a high degree of homology to chicken ovalbumin, lack of N- and C-terminal extensions, absence of a signal peptide, and a serine rather than an asparagine residue at the penultimate positionSpecifications
| SERPINB8 | |
| Polyclonal | |
| Unconjugated | |
| SERPINB8 | |
| CAP 2, CAP2, Cytoplasmic antiproteinase 2, Peptidase inhibitor 8, PI 8, PI8, Serpin B8, SERPINB8 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20725, 5271 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O08800, P50452 | |
| SERPINB8 | |
| SERPINB8 Fusion Protein Ag3740 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction