missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINB6 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518947
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
The protein encoded by this gene is a member of the serpin superfamily, and ovalbumin-serpin subfamily. It was originally discovered as a placental thrombin inhibitor. The mouse homolog was found to be expressed in the hair cells of the inner ear. Mutations in this gene are associated with nonsyndromic progressive hearing loss, suggesting that this serpin plays an important role in the inner ear in the protection against leakage of lysosomal content during stress, and that loss of this protection results in cell death and sensorineural hearing loss. Alternatively spliced transcript variants have been found for this gene.Specifications
SERPINB6 | |
Polyclonal | |
Unconjugated | |
SERPINB6 | |
4930482L21Rik; CAP; Cytoplasmic antiproteinase; D330015H01Rik; DFNB91; DKFZp686I04222; MGC111370; MSTP057; ovalbumin; Peptidase inhibitor 6; PI6; PI-6; Placental thrombin inhibitor; protease inhibitor 6 (placental thrombin inhibitor); proteinase inhibitor 6; PTI; QtsA-17114; RP1-90J20.6; serine (or cysteine) peptidase inhibitor, clade B, member 6a; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 6; serine (or cysteine) proteinase inhibitor, clade B, member 6a; serine protease inhibitor 3; serine proteinase inhibitor B-43; Serpin B6; serpin family B member 6; serpin peptidase inhibitor, clade B (ovalbumin), member 6; Serpinb6; Serpinb6a; Spi3; thrombin inhibitor; unnamed protein product | |
Goat | |
Antigen affinity chromatography | |
RUO | |
5269 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P35237 | |
SERPINB6 | |
Synthetic peptide sequence (NWDEQFDKENTEER) corresponding to the internal amino acids of SERPINB6 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |