missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINB4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550131
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total SERPINB4 protein.
SERPINB4 Polyclonal antibody specifically detects SERPINB4 in Human samples. It is validated for Western Blot.Specifications
SERPINB4 | |
Polyclonal | |
Unconjugated | |
SERPINB4 | |
1110001H02Rik; 1110013A16Rik; LEUPIN; Leupin SERPINB4; Peptidase inhibitor 11; PI11; PI-11; protease inhibitor (leucine-serpin); RGD1562868; SCCA1; SCCA2; SCCA-2; SCCA2/SCCA1 fusion protein; serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 3A; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 3A; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 4; serine peptidase inhibitor, clade B, member 3A; serpin B4; serpin family B member 4; serpin peptidase inhibitor, clade B (ovalbumin), member 4; Serpinb3a; SERPINB4; Sqn5; squamous cell carcinoma antigen 1; squamous cell carcinoma antigen 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6318 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P48594 | |
SERPINB4 | |
Synthesized peptide derived from internal of human SERPINB4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |