missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINB4 Antibody (001), DyLight 680, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90154FR
This item is not returnable.
View return policy
Description
SERPINB4 Monoclonal specifically detects SERPINB4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SERPINB4 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Leupin, Peptidase inhibitor 11, PI-11, PI11clade B (ovalbumin), member 4, serpin peptidase inhibitor, clade B (ovalbumin), member 4, squamous cell carcinoma antigen 1, Squamous cell carcinoma antigen 2 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human SERPINB4 (P48594; Met1-Pro390). | |
| 0.1 mL | |
| Immunology | |
| 6318 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction