missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SERPINB3 Monoclonal Antibody (798CT5.2.2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537718
This item is not returnable.
View return policy
Description
SERPINB3 Monoclonal Antibody for Western Blot
May act as a papain-like cysteine protease inhibitor to modulate the host immune response against tumor cells. Also functions as an inhibitor of UV-induced apoptosis via suppression of the activity of c-Jun NH(2)-terminal kinase (JNK1). [UniProt].
Specifications
| SERPINB3 | |
| Monoclonal | |
| Unconjugated | |
| SERPINB3 | |
| HsT1196; LOC483954; Protein T4-A; SCC; SCCA; SCCA1; SCCA-1; SCCA-PD; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 3; serine protease inhibitor B3; serpin B3; serpin family B member 3; serpin peptidase inhibitor, clade B (ovalbumin), member 3; serpin peptidase inhibitor, clade B (ovalbumin), member 4; SERPINB3; SERPINB4; spb3; Squamous cell carcinoma antigen 1; SSCA1; T4-A | |
| Mouse | |
| Protein G | |
| RUO | |
| 6317 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 798CT5.2.2 | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P29508 | |
| SERPINB3 | |
| recombinant between 1-390 amino acids from human SERPINB3. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction