missing translation for 'onlineSavingsMsg'
Learn More

SERPINB13 Antibody, DyLight 550, Novus Biologicals™

Product Code. 18422887 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18422887 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18422887 Supplier Novus Biologicals Supplier No. NBP224381R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SERPINB13 Polyclonal antibody specifically detects SERPINB13 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERPINB13
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias HaCaT UV-repressible serpin, Headpin, HSHUR7SEQ, HUR7, HURPIN, MGC126870, Peptidase inhibitor 13, PI-13, PI13hurpin, protease inhibitor 13 (hurpin, headpin), Proteinase inhibitor 13, serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 13, serpin B13, serpin peptidase inhibitor, clade B (ovalbumin), member 13, UV-B repressed sequence, HUR 7
Host Species Rabbit
Immunogen Reacts with residues 310-324 [SEHKADYSGMSSGSG] of the human SERPINB13 protein.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Cell Biology, Endocrinology
Primary or Secondary Primary
Gene ID (Entrez) 5275
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.