missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINB10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21719-1-AP-20UL
This item is not returnable.
View return policy
Description
Protease inhibitor that may play a role in the regulation of protease activities during hematopoiesis and apoptosis induced by TNF. May regulate protease activities in the cytoplasm and in the nucleus.Specifications
| SERPINB10 | |
| Polyclonal | |
| Unconjugated | |
| SERPINB10 | |
| bomapin, Peptidase inhibitor 10, PI 10, PI10, Serpin B10, SERPINB10 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 241197, 266775, 5273 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P48595, Q8K1K6, Q8K3K4 | |
| Serpinb10 | |
| SERPINB10 Fusion Protein Ag16372 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction