missing translation for 'onlineSavingsMsg'
Learn More

SERPINB10 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18417306 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18417306 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18417306 Supplier Novus Biologicals Supplier No. NBP224382MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SERPINB10 Polyclonal antibody specifically detects SERPINB10 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERPINB10
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias bomapin, Peptidase inhibitor 10, PI-10, PI10protease inhibitor 10 (ovalbumin type, bomapin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 10, serpin B10, serpin peptidase inhibitor, clade B (ovalbumin), member 10
Host Species Rabbit
Immunogen Reacts with residues 316-330 [SQSKADFSGMSSARN] of the human SERPINB10 protein.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5273
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.