missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINB10 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98324IR
This item is not returnable.
View return policy
Description
SERPINB10 Polyclonal specifically detects SERPINB10 in Mouse samples. It is validated for ELISA.
Specifications
| SERPINB10 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 5273 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| DyLight 755 | |
| bomapin, Peptidase inhibitor 10, PI-10, PI10protease inhibitor 10 (ovalbumin type, bomapin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 10, serpin B10, serpin peptidase inhibitor, clade B (ovalbumin), member 10 | |
| Produced in rabbits immunized with purified, recombinant SERPINB10 (Q8K1K6-1; Met1-Pro397). | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction