missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SERPINA6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA524229
This item is not returnable.
View return policy
Description
SERPINA6 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes an alpha-globulin protein with corticosteroid-binding properties. This is the major transport protein for glucorticoids and progestins in the blood of most vertebrates. The gene localizes to a chromosomal region containing several closely related serine protease inhibitors which may have evolved by duplication events.
Specifications
| SERPINA6 | |
| Polyclonal | |
| Unconjugated | |
| SERPINA6 | |
| AI265318; AV104445; Cbg; corticosteroid binding globulin; corticosteroid-binding globulin; serine (or cysteine) peptidase inhibitor, clade A, member 6; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 6; serine (or cysteine) proteinase inhibitor, clade A, member 6; serpin A6; serpin family A member 6; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 6; serpin peptidase inhibitor, clade A, member 6; Serpina6; Transcortin | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 866 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.27 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P08185 | |
| SERPINA6 | |
| KLH conjugated synthetic peptide between 197-225 amino acids from the central region of human SERPINA6. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction