missing translation for 'onlineSavingsMsg'
Learn More

SERPINA6 293T Cell Overexpression Lysate (Denatured), Abnova

Product Code. 16125724
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 16125724

Brand: Abnova H00000866T01.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Western Blotting

Specifications

Cell Line 293T
For Use With (Application) Western Blot
Description SERPINA6 293T Cell Transient Overexpression Lysate(Denatured)
Gene Accession No. NM_001756.3
Molecular Weight (g/mol) 44.66
pH Range 6.8
Plasmid pCMV-SERPINA6 full-length
Quality Control Testing Transient overexpression cell Lysate was tested with Anti-SERPINA6 antibody (H00000866-B01) by western blotting
Quantity 100 μL
Storage Buffer 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Host Species Human
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.