missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SERPINA3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585287
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Rhesus Monkey (86%). Positive Control: U87-MG, SK-N-SH, IMR32, SK-N-AS, human SERPIN-transfected 293T. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SERPINA encodes Alpha 1-ACT. It is an early-stage acute-phase plasma protein and a serpin that preferentially inactivates chymotrypsin, cathepsin G, and chymase. Alpha-1-ACT, a serine protease inhibitor, is tightly associated with amyloid plaques in Alzheimer's disease (AD) and in normal aged human and monkey brain. Regulation of the serine proteases and serine protease inhibitors plays an important role in neuromuscular differentiation. Prostate specific antigen (PSA), a chymotrypsin-like serine protease, is predominantly complexed to Alpha-1-ACT.
Specifications
| SERPINA3 | |
| Polyclonal | |
| Unconjugated | |
| SERPINA3 | |
| AACT; ACT; alpha-1-antichymotrypsin; Alpha-1-antichymotrypsin His-Pro-less; Cell growth-inhibiting gene 24/25 protein; GIG24; GIG25; Growth inhibiting protein 24; Growth inhibiting protein 25; growth-inhibiting protein 24; growth-inhibiting protein 25; MGC88254; Serine (or cysteine) proteinase inhibitor clade A member 3; serine (or cysteine) proteinase inhibitor, clade A, member 3; serpin A3; serpin family A member 3; Serpin peptidase inhibitor clade A (alpha 1 antiproteinase antitrypsin) member 3; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 3; SERPINA3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P01011 | |
| SERPINA3 | |
| Recombinant protein encompassing a sequence within the center region of human alpha 1 Antichymotrypsin. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction