missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SERPINA3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

£263.00 - £338.00

Specifications

Antigen SERPINA3
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11517262
View Documents
Invitrogen™
PA137070
1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
13496777
GREENER_CHOICE
View Documents
Invitrogen™
PA532260
500 μL
£263.00
500µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

A suggested positive control is tonsil tissue.

SERPINA encodes Alpha 1-ACT. It is an early-stage acute-phase plasma protein and a serpin that preferentially inactivates chymotrypsin, cathepsin G, and chymase. Alpha-1-ACT, a serine protease inhibitor, is tightly associated with amyloid plaques in Alzheimer's disease (AD) and in normal aged human and monkey brain. Regulation of the serine proteases and serine protease inhibitors plays an important role in neuromuscular differentiation. Prostate specific antigen (PSA), a chymotrypsin-like serine protease, is predominantly complexed to Alpha-1-ACT.
Specifications

Specifications

SERPINA3
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
RUO
PBS with 1% BSA and 0.1% sodium azide; pH 7.6
P01011
12
Purified human alpha-1-antichymotrypsin.
Antibody
-20°C, Avoid Freeze/Thaw Cycles
Polyclonal
Liquid
IgG
Human
SERPINA3
AACT; ACT; alpha-1-antichymotrypsin; Alpha-1-antichymotrypsin His-Pro-less; Cell growth-inhibiting gene 24/25 protein; GIG24; GIG25; Growth inhibiting protein 24; Growth inhibiting protein 25; growth-inhibiting protein 24; growth-inhibiting protein 25; MGC88254; Serine (or cysteine) proteinase inhibitor clade A member 3; serine (or cysteine) proteinase inhibitor, clade A, member 3; serpin A3; serpin family A member 3; Serpin peptidase inhibitor clade A (alpha 1 antiproteinase antitrypsin) member 3; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 3; SERPINA3
SERPINA3
Primary
Antigen affinity chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.