missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SERPINA11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106741
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SERPINA11.
The protein belongs to the serpin family. SERPINA11 has serine-type endopeptidase inhibitor activity, negative regulation of endopeptidase activity, regulation of proteolysis, serine-type endopeptidase inhibitor activity.
Specifications
| SERPINA11 | |
| Polyclonal | |
| Unconjugated | |
| SERPINA11 | |
| antiproteinase-like 2; Gm895; serine (or cysteine) peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 11; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 11; serine (or cysteine) proteinase inhibitor, clade A, member 11; serpin A11; serpin family A member 11; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 11; Serpina11 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 256394 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q86U17 | |
| SERPINA11 | |
| A synthesized peptide derived from human SERPINA11(Accession Q86U17), corresponding to amino acid residues W5-T55. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction