Learn More
SERPINA10 Mouse anti-Human, CoraLite 594, Clone: 5B3C9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-60113-100UL
Description
The SERPINA10 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SERPINA10. This antibody recognizes human, rat, pig antigen. The SERPINA10 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| SERPINA10 | |
| Monoclonal | |
| 2000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Pig |
| Immunofluorescence | |
| 5B3C9 | |
| CoraLite 594 | |
| 100 μL | |
| 51156 | |
| IgG2a |