missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SERPINA10 Monoclonal Antibody (C4), Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA550955
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the serpin family. It is predominantly expressed in the liver and secreted in plasma. It inhibits the activity of coagulation factors Xa and XIa in the presence of protein Z, calcium and phospholipid. Mutations in this gene are associated with venous thrombosis. Alternatively spliced transcript variants have been found for this gene.
Specifications
| SERPINA10 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| Q62975, Q9UK55 | |
| SERPINA10 | |
| Recombinant Human SERPINA10 protein, Pro24-Leu444 (Accession #Q9UK55). | |
| 200 μL | |
| Primary | |
| Human, Pig, Rat | |
| Antibody | |
| IgG2a κ |
| Immunohistochemistry (Paraffin), Western Blot | |
| C4 | |
| Unconjugated | |
| SERPINA10 | |
| plasma protein associated with liver regeneration; protein Z-dependent protease inhibitor; PZ-dependent protease inhibitor; PZI; Rasp1; RASP-1; Regeneration-associated serpin 1; serine (or cysteine) peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10; serpin A10; serpin family A member 10; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10; serpin peptidase inhibitor, clade A, member 10; Serpina10; UNQ707/PRO1358; Zpi | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 171154, 51156 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction