missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Serpin I2 293T Cell Transient Overexpression Lysate

Product Code. 18368029 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.01mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368029 0.1 mL 0.01mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18368029 Supplier Novus Biologicals™ Supplier No. H00005276T01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Overexpression Lysate

TRUSTED_SUSTAINABILITY

Specifications

Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.
Target Serpin I2
Synonym MEPIpancpin, Myoepithelium-derived serine protease inhibitor, Pancpin, Pancreas-specific protein TSA2004, Peptidase inhibitor 14, PI-14, PI14clade I (neuroserpin), member 2, protease inhibitor 14, serine (or cysteine) proteinase inhibitor, clade I (pancpin), member 2, serpin I2, serpin peptidase inhibitor, clade I (pancpin), member 2, SERPIN12
Gene Symbol SERPINI2
Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.