missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin I1/Neuroserpin Antibody (002), mFluor Violet 610 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Serpin I1/Neuroserpin Monoclonal antibody specifically detects Serpin I1/Neuroserpin in Mouse samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Serpin I1/Neuroserpin |
| Classification | Monoclonal |
| Dilution | Immunohistochemistry-Paraffin |
| Gene Alias | DKFZp781N13156, neuroserpin, Peptidase inhibitor 12, PI-12, PI12Serpin I1, serine (or cysteine) proteinase inhibitor, clade I (neuroserpin), member 1, serpin peptidase inhibitor, clade I (neuroserpin), member 1 |
| Purification Method | Protein A purified |
| Regulatory Status | RUO |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?