missing translation for 'onlineSavingsMsg'
Learn More

Serpin G1/C1 Inhibitor Antibody, FITC, Novus Biologicals™

Product Code. 18342682 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18342682 0.1 mL 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18342682 Supplier Novus Biologicals Supplier No. NBP311027F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Serpin G1/C1 Inhibitor Polyclonal antibody specifically detects Serpin G1/C1 Inhibitor in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serpin G1/C1 Inhibitor
Applications ELISA
Classification Polyclonal
Conjugate FITC
Dilution ELISA
Formulation PBS
Gene Alias C1IN, C1INH, C1NH, HAE1, HAE2, SERPING1 serpin peptidase inhibitor, clade G (C1 inhibitor), member 1
Host Species Sheep
Immunogen Human Serpin G1/C1 Inhibitor
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 710
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.